Learn More
IGF1R beta Monoclonal Antibody (3G5C1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515354
Code : A0
Additional Details : Weight : 0.01000kg
Description
MA5-15354 targets IGF1R in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15354 immunogen is purified recombinant fragment of IGF1R-Beta (Amino Acid: 1101-1367) expressed in E. Coli.
IGF1R beta Monoclonal antibody specifically detects IGF1R beta in Human samples. It is validated for ELISA, Immunohistochemistry, Western Blot.Specifications
IGF1R beta | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P08069 | |
IGF1R | |
Purified recombinant fragment of IGF1R-Beta (Amino Acid: 1101-1367) expressed in E. Coli. | |
RUO | |
3480 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
3G5C1 | |
Unconjugated | |
IGF1R | |
A330103N21Rik; CD221; D930020L01; hyft; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |