missing translation for 'onlineSavingsMsg'
Learn More

IFN gamma, Oligoclonal, Invitrogen™

Rabbit Oligoclonal Antibody

Brand:  Invitrogen 710152

Code : NEW

Additional Details : Weight : 0.00450kg

Product Code. 15387214

  • £281.00 / 100µg


EU Stock 1
Add to basket

Description

Description

ABfinity™ recombinant antibodies are rabbit monoclonal antibodies, unmatched for producing superior results. ABfinity™ antibodies are developed by immunizing animals, screening for functionality, cloning the immunogen-specific antibody genes into high-level mammalian expression vectors, produced on a large scale and purified with Protein A. ABfinity™ oligoclonal antibodies comprise a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds—the sensitivity of a polyclonal antibody with the specificity of a monoclonal, all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody—recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets when compared with monoclonal antibodies—an oligoclonal antibody has a known mixture of light and heavy chains. This exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.Intact IgG appears on a non-reducing gel as ∽150kDa band and upon reduction generating a ∽25kDa light chain band and a ∽50kDa heavy chain.
Specifications

Specifications

IFN gamma
Oligoclonal
0.5mg/mL
PBS with 0.09% sodium azide
P01581
IFN-gamma, boIFNG, interferon gamma, interferon gamma type 2, interferon-gamma, IFNG, IFI, IFG
Rabbit
IgG
100μg
Antibody
Oligoclonal
Rat
Western Blot
18HCLC
Unconjugated
Ifng
Purified
IFNG
Recombinant protein corresponding to amino acids 23–156 of rat interferon gamma
Protein A
Store at 2-8°C for up to 1 month. For long term storage store at -20°C
Primary
25712
Videos