Learn More
IFN gamma Monoclonal Antibody (B133.5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen M701
Code : PN
Additional Details : Weight : 0.80000kg
Description
M701 was successfully to detect IFN-gamma in Flow application. M701 targets IFN gamma in Western blot applications and shows reactivity with Human samples. The M700A anti-IFN Gamma antibody (clone 2G1) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M701B (biotinylated conjugate of clone B133.5). Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. The M701 immunogen is recombinant human IFN gamma. M701 detects IFN gamma which has a predicted molecular weight of approximately 16 kDa. This product has been tested for endotoxins by limulus amoebocyte lysate (LAL) assay and contains an endotoxin concentration of less than or equal to 10 endotoxin units per milligram (EU/mg).
IFN gamma Monoclonal antibody specifically detects IFN gamma in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
IFN gamma | |
Monoclonal | |
1.0 mg/mL | |
PBS with no preservative | |
P01579 | |
IFNG | |
Recombinant human IFN gamma | |
500 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry, Western Blot | |
B133.5 | |
Unconjugated | |
IFNG | |
BoIFNG; cytokine; EGK_03901; gamma interferon; gamma interferon precursor; gamma-interferon; gamma-interferon precursor (AA -23 to 143); H-IFN-g; IFG; IFI; IFN gamma; IFN ?; Ifng; IFN-g; IFNG2; IFN-gamma; IFN-gamma precursor; IFN-y; IFN?; Immune interferon; interferon gamma; interferon gamma precursor; interferon gamma type 2; Interferon y; Interferon ?; interferon, gamma; interferon-gama; interferon-gamma; Interferon-gamma level; interferon-gamma precursor; Interferon?; M-IFN-g; R-IFN-g | |
Mouse | |
Protein A | |
RUO | |
3458 | |
-20°C | |
Liquid |