missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IDH1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA519414
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, canine and ovine based on sequence homology.
IDH1 Polyclonal antibody specifically detects IDH1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
IDH1 | |
Polyclonal | |
Unconjugated | |
IDH1 | |
AI314845; AI788952; cb876; corneal epithelial crystallin; cytosolic NADP+-dependent isocitrate dehydrogenase; cytosolic NADP-isocitrate dehydrogenase; cytosolic NADP-isocitrate dehydrongenase; E030024J03Rik; epididymis luminal protein 216; epididymis secretory protein Li 26; fm90e09; HEL-216; HEL-S-26; ICDH; Id-1; IDCD; IDH; Idh1; Idh-1; IDP; Idpc; im:7143416; isocitrate dehydrogenase (NAD(+)) IDH1; isocitrate dehydrogenase (NADP(+)) 1; isocitrate dehydrogenase (NADP(+)) 1, cytosolic; Isocitrate dehydrogenase [NAD] subunit 1, mitochondrial; Isocitrate dehydrogenase [NADP] cytoplasmic; isocitrate dehydrogenase 1 (NADP+); isocitrate dehydrogenase 1 (NADP+), soluble; Isocitrate dehydrogenase 1, soluble; Isocitric dehydrogenase; N2690; NAD(+)-specific ICDH; NADP(+)-specific ICDH; NADP+-specific ICDH; NADP-cICDH; NADP-dependent isocitrate dehydrogenase, cytosolic; NADP-dependent isocitrate dehydrogenase, peroxisomal; OTTHUMP00000206464; OTTHUMP00000206465; OTTHUMP00000206466; oxalosuccinate | |
Goat | |
Antigen affinity chromatography | |
RUO | |
15926, 24479, 3417 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
O75874, O88844, P41562 | |
IDH1 | |
Synthetic peptide sequence (KKHNVGVKCATIT) corresponding to the internal amino acids of IDH1 | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |