missing translation for 'onlineSavingsMsg'
Learn More

ICAM-5 Mouse anti-Human, Clone: 283607, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1950

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459055

  • £271.93 / 100µg


Estimated Shipment
29-11-2018
Add to basket

Description

Description

ICAM-5 Monoclonal antibody MAB1950 Specifically detects ICAM-5 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications

Specifications

ICAM-5
283607
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
ICAM5
Mouse myeloma cell line NS0-derived recombinant human ICAM-5, Ala28-Glu570, Accession # Q9UMF0
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7087
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Immunocytochemistry
LYOPH
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL
Q9UMF0
ICAM5, ICAM-5, intercellular adhesion molecule 5, intercellular adhesion molecule 5, telencephalin, Telencephalin, TLCNTelencephalin, TLNtelencephalin
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human ICAM-5 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) CD31, rhCEACAM-1, recombinant mouse (rm) DCC, rhICAM-1, -2, -3, rmICAM-5, rhMAdCAM-1, or rmVCAM-1 is observed.
Human
Videos

For Research Use Only