missing translation for 'onlineSavingsMsg'
Learn More
Learn More
I-Ab/I-Ad Monoclonal Antibody (25-9-17S), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA517775
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17775 targets I-Ab/I-Ad in FACS applications and shows reactivity with Mouse samples. The MA5-17775 immunogen is c3H.SW splenocytes.
I-Ab/I-Ad Monoclonal antibody specifically detects I-Ab/I-Ad in Mouse samples. It is validated for Flow Cytometry.Specifications
I-Ab/I-Ad | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P01921, P06342, P06343, P06344, P06345, P06346, P14483 | |
H2-Ab1 | |
C3H.SW splenocytes | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a |
Flow Cytometry | |
25-9-17S | |
Biotin | |
H2-Ab1 | |
Abeta; AI845868; H-2 class II histocompatibility antigen, A beta chain; H-2Ab; H2-Ab; H2-Ab1; H2-iabeta; histocompatibility 2, class II antigen A, beta 1; Ia2; Ia-2; IAb; I-Abeta; major histocompatibility complex class II beta chain; MHC class II antigen A beta; MHC class II H2-IA-beta-psi; response to metastatic cancers 1; Rmcs1 | |
Mouse | |
Protein G | |
RUO | |
14961 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |