missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HUNK Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533857
Code : NEW
Additional Details : Weight : 0.01000kg
Description
HUNK Polyclonal antibody specifically detects HUNK in Bovine, Canine, Chicken, Equine, Hamster, Human, Mouse, Non-human Primate, Rat samples. It is validated for Immunohistochemistry (Paraffin).Specifications
HUNK | |
Polyclonal | |
Unconjugated | |
HUNK | |
B19; Bstk1; Hormonally up-regulated neu tumor-associated kinase; hormonally upregulated neu tumor-associated kinase; hormonally up-regulated Neu-associated kinase; hormonally upregulated Neu-associated kinase; Hunk; MAKV; Mak-v; RGD1565091; serine/threonine protein kinase MAK-V; serine/threonine-protein kinase MAK-V | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100064062, 100765960, 26559, 288275, 30811, 418493, 487733, 537640 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O88866, P57058 | |
HUNK | |
Synthetic 18 amino acid peptide from Protein kinase domain of human HUNK. | |
50 μg | |
Primary | |
Bovine, Canine, Chicken, Equine, Hamster, Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |