missing translation for 'onlineSavingsMsg'
Learn More

Wnt-9a Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3157

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438014

  • £314.17 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Wnt-9a Polyclonal antibody AF3157 Specifically detects Wnt-9a in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Wnt-9a
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
O14904.2
MGC138165, MGC141991, Protein Wnt-14, wingless-type MMTV integration site family, member 14, wingless-type MMTV integration site family, member 9A, Wnt-14, WNT14protein Wnt-9a, Wnt9a
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Wnt-9a in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant mouse (rm) Wnt-9b is observed and less than 1% cross-reactivity with rmWnt-1 and rmWnt-4 is observed.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
WNT9A
E. coli-derived recombinant human Wnt-9a Pro48-Glu92 & Arg231-Phe304 Accession # O14904.2
Immunogen affinity purified
RUO
Primary
7483
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only