missing translation for 'onlineSavingsMsg'
Learn More

VSIG4 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4646

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448834

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

VSIG4 Polyclonal antibody AF4646 Specifically detects VSIG4 in Human samples. It is validated for Western Blotting
Specifications

Specifications

VSIG4
LYOPH
Western Blot 0.1 ug/mL
Q9Y279
CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IG, Z39IGcomplement receptor of the immunoglobulin superfamily
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human VSIG4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 15% cross-reactivity with recombinant mouse VSIG4 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
VSIG4
Mouse myeloma cell line NS0-derived recombinant human VSIG4 Arg20-Pro283 Accession # Q9Y279
Immunogen affinity purified
RUO
Primary
11326
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only