missing translation for 'onlineSavingsMsg'
Learn More

VEGF-B 167/186 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF751

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13450135

  • £464.72 / 100µg

Estimated Shipment
15-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

VEGF-B 167/186 Polyclonal antibody AF751 Specifically detects VEGF-B 167/186 in Human samples. It is validated for Western Blotting
Specifications

Specifications

VEGF-B
LYOPH
Western Blot 0.1 ug/mL
Affinity Purified
VEGFB
E. coli-derived recombinant human VEGF-B 186
Immunogen affinity purified
RUO
Primary
7423
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
vascular endothelial growth factor B, VEGFB, VEGF-B, VEGF-related factor, VRFVEGFL
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human VEGF-B 167 and human VEGF-B 186 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) VEGF-B 186, rmVEGF-B 167, rmVEGF-B 206, recombinant human (rh) VEGF-A, rhVEGF-C, and rhVEGF-D is observed.
Human
Videos
Documents

For Research Use Only