missing translation for 'onlineSavingsMsg'
Learn More

Vasohibin Isoform 1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3319

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438104

  • £449.92 / 100µg

Estimated Shipment
10-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Vasohibin Isoform 1 Polyclonal antibody AF3319 Specifically detects Vasohibin Isoform 1 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Vasohibin
LYOPH
Western Blot 0.1 ug/mL
Q7L8A9
KIAA1036, KIAA1036vasohibin-1, VASH, VASH1, vasohibin 1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Vasohibin Isoform 1 in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
VASH1
Mouse myeloma cell line NS0-derived recombinant human Vasohibin Isoform 1 Pro2-Val365 Accession # Q7L8A9
Immunogen affinity purified
RUO
Primary
22846
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only