missing translation for 'onlineSavingsMsg'
Learn More

Tryptase alpha/TPS1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2370

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437514

  • £398.37 / 100µg

Estimated Shipment date
from Supplier 25-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Tryptase alpha/TPS1 Polyclonal antibody AF2370 Specifically detects Tryptase alpha/TPS1 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Tryptase alpha/TPS1
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
AAA86934
TPS1, TPS2, TPSAB1, TPSB1, Tryptase 1, tryptase alpha/beta 1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Tryptase alpha/TPS1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 60% cross-reactivity with recombinant human (rh) TPSB2 is observed and 10% cross-reactivity with rhTPSG1 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TPSAB1
Mouse myeloma cell line NS0-derived recombinant human Tryptase alpha/TPS1 Ile31-Pro275 Accession # AAA86934
Immunogen affinity purified
RUO
Primary
7177
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only