missing translation for 'onlineSavingsMsg'
Learn More

TROY/TNFRSF19 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1548

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437004

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

TROY/TNFRSF19 Polyclonal antibody AF1548 Specifically detects TROY/TNFRSF19 in Human samples. It is validated for Western Blotting
Specifications

Specifications

TROY/TNFRSF19
LYOPH
Western Blot 0.1 ug/mL
AAF71828
TAJ, TAJ-alpha, TAJToxicity and JNK inducer, TNFRSF19, TRADE, TROYtumor necrosis factor receptor superfamily member 19, tumor necrosis factor receptor superfamily, member 19
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human TROY/TNFRSF19 in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) TROY is observed and less than 1% cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhCD30, rhCD40, rhDR3, rhDR6, rhEDAR, rhFas, rhGITR, rhHVEM, rhNGF R, rhOPG, rhRANK, rhTNF RI, and rhTNF RII is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
TNFRSF19
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human TROY/TNFRSF19 Glu30-Leu170 Accession # AAF71828
Immunogen affinity purified
RUO
Primary
55504
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only