Learn More
Thymopoietin Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF843
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13460265
  • £314.17 / 100µg
Estimated Shipment
28-02-2019
Add to basket
Description
Description
Thymopoietin Polyclonal antibody AF843 Specifically detects Thymopoietin in Human samples. It is validated for Western Blotting
Specifications
Specifications
Thymopoietin/LAP2
LYOPH
Western Blot 0.1 ug/mL
Affinity Purified
TMPO
E. coli-derived recombinant human Thymopoietin Pro2-Gln187
Immunogen affinity purified
RUO
Primary
7112
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CMD1T, lamina-associated polypeptide 2, LAP2PRO0868, LEM domain containing 4, LEMD4, MGC61508, thymopoietin, Thymopoietin isoform alpha, Thymopoietin, isoforms beta/gamma, Thymopoietin-related peptide isoform alpha, Thymopoietin-related peptide isoforms beta/gamma, TMPO, TP, TP alpha, TP beta/gamma, TPRP isoform alpha, TPRP isoforms beta/gamma
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Thymopoietin in direct ELISAs and Western blots.