missing translation for 'onlineSavingsMsg'
Learn More

Thrombospondin-2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1635

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437064

  • £449.92 / 100µg

Estimated Shipment
13-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Thrombospondin-2 Polyclonal antibody AF1635 Specifically detects Thrombospondin-2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Thrombospondin-2
LYOPH
Western Blot 0.1 ug/mL
P35442
THBS2, thrombospondin 2, Thrombospondin2, TSP-2, TSP2thrombospondin-2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Thrombospondin-2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Thrombospondin-1 and rhThrombospondin-4 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
THBS2
Mouse myeloma cell line NS0-derived recombinant human Thrombospondin-2 Gly19-Ile1172 Accession # P35442
Immunogen affinity purified
RUO
Primary
7058
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only