missing translation for 'onlineSavingsMsg'
Learn More

TGF-beta RII Isoform 2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1003

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13486654

  • £382.00 / 100µg


Estimated Shipment
05-07-2018
Add to basket

Description

Description

TGF-beta RII Isoform 2 Polyclonal antibody AF1003 Specifically detects TGF-beta RII Isoform 2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

TGF-beta RII Isoform 2
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant human TGF-β RII Isoform 2, Thr23-Asp184, Accession No. NP_001020018
100μg
Primary
7048
Unconjugated
NP_001020018
Goat
IgG
RUO
Polyclonal
Human
Videos

For Research Use Only