missing translation for 'onlineSavingsMsg'
Learn More

Syndecan-3 Isoform 1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3539

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13428224

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Syndecan-3 Isoform 1 Polyclonal antibody AF3539 Specifically detects Syndecan-3 Isoform 1 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blotting
Specifications

Specifications

SDC3
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
O75056
KIAA0468, N-syndecan, SDC3, SDCN, SYND3syndecan proteoglycan 3, syndecan 3, syndecan 3 (N-syndecan), syndecan neural type, syndecan-3, Syndecan3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Syndecan-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse Syndecan-3 is observed and less than 1% cross-reactivity with recombinant mouse Syndecan-4 and recombinant human Syndecan-2 is observed.
Human
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SDC3
Mouse myeloma cell line NS0-derived recombinant human Syndecan-3 Gln48-Lys383 Accession # O75056
Immunogen affinity purified
RUO
Primary
9672
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only