missing translation for 'onlineSavingsMsg'
Learn More

Stanniocalcin 2/STC-2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2830

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447814

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 31-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Stanniocalcin 2/STC-2 Polyclonal antibody AF2830 Specifically detects Stanniocalcin 2/STC-2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Stanniocalcin 2/STC-2
LYOPH
Western Blot 0.1 ug/mL
O76061
stanniocalcin 2, stanniocalcin-2, STC2, STC-2, STC-2Stanniocalcin-related protein, STC-related protein, STCRP
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human and mouse Stanniocalcin 2/STC-2 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human STC-1 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
STC2
Mouse myeloma cell line NS0-derived recombinant human Stanniocalcin 2/STC-2 Thr25-Arg302 Accession # O76061
Antigen Affinity-purified
RUO
Primary
8614
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only