missing translation for 'onlineSavingsMsg'
Learn More

Stanniocalcin 1/STC-1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2958

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457894

  • £389.41 / 100µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Stanniocalcin 1/STC-1 Polyclonal antibody AF2958 Specifically detects Stanniocalcin 1/STC-1 in Human samples. It is validated for Western Blotting
Specifications

Specifications

Stanniocalcin 1/STC-1
LYOPH
Western Blot 0.1 ug/mL
P52823
stanniocalcin 1, stanniocalcin-1, STC1, STC-1, STCSTC-1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Stanniocalcin 1/STC-1 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant human STC-2 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
STC1
Mouse myeloma cell line NS0-derived recombinant human Stanniocalcin 1 Thr18-Ala247 Accession # P52823
Immunogen affinity purified
RUO
Primary
6781
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only