missing translation for 'onlineSavingsMsg'
Learn More

SOX7 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2766

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13427774

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SOX7 Polyclonal antibody AF2766 Specifically detects SOX7 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

SOX7
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q9BT81
MGC10895, SOX7 transcription factor, SRY (sex determining region Y)-box 7, transcription factor SOX-7
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human SOX7 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant human SOX1, -2, -3, -9, -10, and -17 is observed.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SOX7
E. coli-derived recombinant human SOX7 Ser249-Ala345 Accession # Q9BT81
Antigen Affinity-purified
RUO
Primary
83595
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only