missing translation for 'onlineSavingsMsg'
Learn More

Smad8 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2309

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447474

  • £332.22 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Smad8 Polyclonal antibody AF2309 Specifically detects Smad8 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Smad8
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
NP_005896
SMAD9
E. coli-derived recombinant human Smad8 Ala158-Ser233 Accession # NP_005896
Immunogen affinity purified
RUO
Primary
4093
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Smad8 in direct ELISAs and Western blots. In direct ELISAs, approximately 90% cross-reactivity with recombinant (rh) Smad1 is observed, approximately 20% cross-reactivity with rhSmad4 is observed, and less than 1% cross-reactivity with rhSmad5, rhSmad7, and rhDNAK is observed.
Human
Videos
Documents

For Research Use Only