missing translation for 'onlineSavingsMsg'
Learn More

SLITRK2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2657

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457704

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 02-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SLITRK2 Polyclonal antibody AF2657 Specifically detects SLITRK2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

SLITRK2
LYOPH
Western Blot 0.1 ug/mL
Q9H156
CXorf2, SLITL1, SLITRK2 SLIT and NTRK-like family, member 2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human SLITRK2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) SLITRK4 is observed and less than 2% cross-reactivity with rhSLITRK1 and rhSLITRK5 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SLITRK2
Mouse myeloma cell line NS0-derived recombinant human SLITRK2 Thr19-Pro617 Accession # Q9H156
Immunogen affinity purified
RUO
Primary
84631
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only