missing translation for 'onlineSavingsMsg'
Learn More

SLITRK1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3009

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437924

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SLITRK1 Polyclonal antibody AF3009 Specifically detects SLITRK1 in Human samples. It is validated for Western Blotting
Specifications

Specifications

SLITRK1
LYOPH
Western Blot 0.1 ug/mL
Q96PX8
FLJ54428, KIAA1910KIAA0918, leucine rich repeat containing 12, Leucine-rich repeat-containing protein 12, LRRC12, RP11-395N17.1, SLIT and NTRK-like family, member 1, SLIT and NTRK-like protein 1, slit and trk like gene 1, TTM
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human SLITRK1 in direct ELISAs and Western blots. In these formats, approximately 5% cross-reactivity with recombinant human (rh) SLITRK2, rhSLITRK4, and rhSLITRK5 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SLITRK1
Mouse myeloma cell line NS0-derived recombinant human SLITRK1 (R&D Systems, Catalog # 3009-SK) Ala16-Ser616 Accession # Q96PX8
Immunogen affinity purified
RUO
Primary
114798
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only