missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serum Albumin Mouse anti-Human, Clone: 1G2, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen LFMA0106
Additional Details : Weight : 0.10000kg
Description
A suggested positive control for this product is human plasma.
Albumin is a soluble, monomeric protein with a molecular weight of approximately 65-67 kDa that comprises about one-half of the blood serum protein. Human serum albumin (HAS) is the most abundant protein in human blood plasma, and has a serum half-life of approximately 20 days. Albumin functions primarily as a carrier protein for steroids, fatty acids, and thyroid hormones and plays a role in stabilizing extracellular fluid volume. Albumin is a globular, unglycosylated serum protein synthesized in the liver as pre-pro-albumin which has an N-terminal peptide that is removed before the nascent protein is released from the rough `produce the secreted albumin. Serum albumin has a good binding capacity for water, Ca(2+), Na(+), K(+), fatty acids, hormones, bilirubin and drugs. The main function of albumin is the regulation of the colloidal osmotic pressure of blood.Specifications
Human Serum Albumin | |
Monoclonal | |
0.1 mg/mL | |
ALB | |
Albumin, HSA, Serum albumin precursor, ALB | |
Mouse | |
Protein G | |
RUO | |
213 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
1G2 | |
Unconjugated | |
P02768 | |
ALB | |
Purified, human serum albumin protein. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 κ |