Learn More
Serpin E2/PN1 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2980
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487904
  • £382.00 / 100µg
Estimated Shipment
01-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
Serpin E2/PN1 Polyclonal antibody AF2980 Specifically detects Serpin E2/PN1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
Serpin E2/PN1
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
Mouse myeloma cell line NS0-derived recombinant human Serpin E2, Met1-Pro397, Accession No. NP_001130000
100μg
Primary
5270
Unconjugated
NP_001130000
Goat