missing translation for 'onlineSavingsMsg'
Learn More

Serpin A12 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4410

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468714

  • £454.23 / 100µg

Estimated Shipment date
from Supplier 25-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Serpin A12 Polyclonal antibody AF4410 Specifically detects Serpin A12 in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Serpin A12
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q8IW75
antitrypsin), member 12, member 12, OL-64, OL-64Visceral adipose-specific serpin, serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, serpin A12, serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), Vaspin, Visceral adipose tissue-derived serine protease inhibitor
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Serpin A12 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) Serpin A1, rhSerpin A3, rhSerpin F1, and recombinant mouse Serpin E1 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SERPINA12
Mouse myeloma cell line NS0-derived recombinant human Serpin A12 Leu20-Lys414 Accession # Q8IW75
Immunogen affinity purified
RUO
Primary
145264
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only