missing translation for 'onlineSavingsMsg'
Learn More

Semaphorin 6D Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2095

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457344

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Semaphorin 6D Polyclonal antibody AF2095 Specifically detects Semaphorin 6D in Human samples. It is validated for Western Blotting
Specifications

Specifications

Semaphorin 6D
LYOPH
Western Blot 0.1 ug/mL
Q8NFY4
KIAA1479FLJ11598, sema domain, transmembrane domain (TM), and cytoplasmic domain, (semaphorin) 6D, Sema6D, semaphorin-6D
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Semaphorin 6D in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) Semaphorin 6C is observed, 5% cross-reactivity with rhSemaphorin 6B is observed and less than 1% cross-reactivity with rhSemaphorin 3A, rhSemaphorin 3B, recombinant mouse (rm) Semaphorin 3E, rmSemaphorin 3F, rhSemaphorin 3A, and rhSemaphorin 7A is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SEMA6D
Mouse myeloma cell line NS0-derived recombinant human Semaphorin 6D Ser22-His660 Accession # Q8NFY4
Immunogen affinity purified
RUO
Primary
80031
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only