missing translation for 'onlineSavingsMsg'
Learn More

SDNSF/MCFD2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2357

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447504

  • £349.86 / 100µg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SDNSF/MCFD2 Polyclonal antibody AF2357 Specifically detects SDNSF/MCFD2 in Human samples. It is validated for Western Blotting
Specifications

Specifications

SDNSF/MCFD2
LYOPH
Western Blot 0.1 ug/mL
Q8NI22
DKFZp686G21263, F5F8D2, LMAN1IP, MCFD2, multiple coagulation factor deficiency 2, multiple coagulation factor deficiency protein 2, neural stem cell derived neuronal survival protein, Neural stem cell-derived neuronal survival protein, SDNSFF5F8D
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human SDNSF/MCFD2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 35% cross-reactivity with recombinant mouse SDNSF is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
MCFD2
E. coli-derived recombinant human SDNSF/MCFD2 Glu27-Gln146 Accession # Q8NI22
Immunogen affinity purified
RUO
Primary
90411
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only