Learn More
SCGF/CLEC11a Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1904
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13487214
  • £352.00 / 100µg
Estimated Shipment
07-03-2018
Add to basket
Disclaimers:
For Research Use Only
Description
Description
SCGF/CLEC11a Polyclonal antibody AF1904 Specifically detects SCGF/CLEC11a in Human samples. It is validated for Western Blotting
Specifications
Specifications
SCGF/CLEC11a
Lyophilized from a 0.2μm filtered solution in PBS with Trehalose.
Affinity Purified
E. coli-derived recombinant human SCGF-β, Gly19-Phe245, Accession No. BAA21499
100μg
Primary
6320
Unconjugated
BAA21499
Sheep