missing translation for 'onlineSavingsMsg'
Learn More

S100A7 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4475

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458744

  • £377.53 / 100µg


Estimated Shipment
04-10-2018
Add to basket

Description

Description

S100A7 Polyclonal antibody AF4475 Specifically detects S100A7 in Human samples. It is validated for Western Blotting
Specifications

Specifications

S100A7/Psoriasin
LYOPH
Western Blot 0.1 ug/mL
AAA60210
HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human S100A7 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human S100A8 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
S100A7
E. coli-derived recombinant human S100A7, Met1-Gln101, Accession # AAA60210
Immunogen affinity purified
RUO
Primary
6278
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only