missing translation for 'onlineSavingsMsg'
Learn More

ROBO2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3147

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448004

  • £449.92 / 100µg

Estimated Shipment date
from Supplier 10-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ROBO2 Polyclonal antibody AF3147 Specifically detects ROBO2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blotting
Specifications

Specifications

ROBO2
LYOPH
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
Q9HCK4
Drosophila) homolog 2, roundabout, axon guidance receptor, homolog 2 (Drosophila), SAX3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human ROBO2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 2% cross-reactivity with recombinant rat ROBO1, recombinant human (rh) ROBO3 and rhROBO4 is observed.
Human
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
ROBO2
Mouse myeloma cell line NS0-derived recombinant human ROBO2 Ser22-Ala312, RPVA, Pro313-Pro859 Accession # Q9HCK4
Immunogen affinity purified
RUO
Primary
6092
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only