missing translation for 'onlineSavingsMsg'
Learn More

RELT/TNFRSF19L Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF1385

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13436914

  • £449.92 / 100µg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

RELT/TNFRSF19L Polyclonal antibody AF1385 Specifically detects RELT/TNFRSF19L in Human samples. It is validated for Western Blotting
Specifications

Specifications

RELT/TNFRSF19L
LYOPH
Western Blot 1 ug/mL
Q969Z4
Receptor expressed in lymphoid tissues, RELT tumor necrosis factor receptor, TNFRSF19L, TNFRSF19LFLJ14993, tumor necrosis factor receptor superfamily member 19L, tumor necrosis factor receptor superfamily, member 19-like
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human RELT/TNFRSF19L in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) 4-1BB, rhBAFF R, rhCD27, rhTAJ, rhCD30, rhDR3, rhDR6, rhTNF RI, rhTNF RII, rhEDAR, rhFas, rhGITR, rhHVEM, rhNGF R, rhOPG, and recombinant mouse OX40 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
RELT
Mouse myeloma cell line NS0-derived recombinant human RELT/TNFRSF19L Ser26-Ala160 (Arg127Gly, Arg129Gly) Accession # Q969Z4
Antigen Affinity-purified
RUO
Primary
84957
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only