missing translation for 'onlineSavingsMsg'
Learn More

Reg1B Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2090

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437344

  • £352.24 / 100µg

Estimated Shipment
13-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Reg1B Polyclonal antibody AF2090 Specifically detects Reg1B in Human samples. It is validated for Western Blotting
Specifications

Specifications

Reg1B
LYOPH
Western Blot 0.1 ug/mL
P48304
Lithostathine 1 beta, lithostathine-1-beta, Pancreatic stone protein 2, pancreatic threadprotein), PSPS2, PSPS2PSP-2, regenerating islet-derived 1 beta, Regenerating islet-derived protein 1-beta, Regenerating protein I beta, REGH, REGLREG-1-beta, secretory pancreatic stone protein 2
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Reg1B in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant mouse (rm) Reg1 is observed, 5% cross-reactivity with rmReg2 and recombinant rat Reg1 is observed and less than 1% cross-reactivity with recombinant human Reg4 and rmReg3A is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
REG1B
E. coli-derived recombinant human Reg1B Gln23-Asn166 Accession # P48304
Immunogen affinity purified
RUO
Primary
5968
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only