missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 3 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF35001

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468204

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 16-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

R-Spondin 3 Polyclonal antibody AF35001 Specifically detects R-Spondin 3 in Human samples. It is validated for Neutralization
Specifications

Specifications

R-Spondin 3
LYOPH
Neutralization 0.3-1.2 ug/mL
Q9BXY4
Cristin 1, CRISTIN1, FLJ14440, hPWTSR, hRspo3, Protein with TSP type-1 repeat, PWTSR, Roof plate-specific spondin-3, RSPO3, RSpondin 3, R-spondin 3 homolog (Xenopus laevis), R-spondin-3, Thrombospondin type-1 domain-containing protein 2, thrombospondin, type I, domain containing 2, THSD2
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human R-Spondin 3 in direct ELISAs. In direct ELISAs, less than 10% cross-reactivity with recombinant mouse R-Spondin 3 is observed.
Human
Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
RSPO3
Chinese hamster ovary cell line CHO-derived recombinant human R-Spondin 3 Gln22-His272 Accession # Q9BXY4
Antigen Affinity-purified
RUO
Primary
84870
Sterile PBS to a final concentration of 0.2 mg/mL.
Videos
Documents

For Research Use Only