missing translation for 'onlineSavingsMsg'
Learn More

R-Spondin 1 Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4645

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438834

  • £314.17 / 100µg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

R-Spondin 1 Polyclonal antibody AF4645 Specifically detects R-Spondin 1 in Human samples. It is validated for Neutralization
Specifications

Specifications

R-Spondin 1
LYOPH
Neutralization 3-12 ug/mL
Q2MKA7
Cristin 3, CRISTIN3, FLJ40906Roof plate-specific spondin-1, hRspo1, roof plate-specific spondin, RSPO, RSPO1, RSPONDIN, RSpondin 1, R-spondin homolog (Xenopus laevis), R-spondin1, R-spondin-1
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human R-Spondin 1 in direct ELISAs. In direct ELISAs, approximately 6% cross-reactivity with recombinant mouse R-Spondin 1 is observed and less than 1% cross-reactivity with recombinant human (rh) R-Spondin 2, rhR-Spondin 3, and rhR-Spondin 4 is observed.
Human
Neutralization
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
RSPO1
Chinese hamster ovary cell line CHO-derived human R-Spondin 1 Arg31-Ala263 Accession # Q2MKA7
Antigen Affinity-purified
RUO
Primary
284654
Sterile PBS to a final concentration of 0.2 mg/mL.
Videos

For Research Use Only