missing translation for 'onlineSavingsMsg'
Learn More

Prolyl Oligopeptidase/PREP Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4308

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13438664

  • £314.17 / 100µg

Estimated Shipment
08-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Prolyl Oligopeptidase/PREP Polyclonal antibody AF4308 Specifically detects Prolyl Oligopeptidase/PREP in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

Prolyl Oligopeptidase/PREP
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
P48147
dJ355L5.1 (prolyl endopeptidase), EC 3.4.21.26, MGC16060, PE, PEP, Post-proline cleaving enzyme, PREP, prolyl endopeptidase, prolyl oligopeptidase, rPop
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human PREP in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 25% cross-reactivity with recombinant mouse PREP is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PREP
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human PREP Leu2-Pro710 Accession # P48147
Antigen Affinity-purified
RUO
Primary
5550
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only