Learn More
PLUNC Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF1897
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13447214
  • £349.86 / 100µg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
PLUNC Polyclonal antibody AF1897 Specifically detects PLUNC in Human samples. It is validated for Western Blotting
Specifications
Specifications
PLUNC
LYOPH
Western Blot 0.1 ug/mL, ELISA
Q9NP55
bA49G10.5, BPIFA1, ligand-binding protein RYA3, LPLUNC3, Lung-specific protein X, LUNX, LUNXNASG, Nasopharyngeal carcinoma-related protein, Palate lung and nasal epithelium clone protein, palate, lung and nasal epithelium associated, palate, lung and nasal epithelium carcinoma associated, protein Plunc, Secretory protein in upper respiratory tracts, SPLUNC1, SPLUNC1SPURT, tracheal epithelium enriched protein, Tracheal epithelium-enriched protein, Von Ebner protein Hl
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human PLUNC in direct ELISAs and Western blots.
Human
Western Blot, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
BPIFA1
E. coli-derived recombinant human PLUNC Gln20-Val256 Accession # Q9NP55
Antigen Affinity-purified
RUO
Primary
51297
Reconstitute at 0.2 mg/mL in sterile PBS.