missing translation for 'onlineSavingsMsg'
Learn More

PLA2G2A Sheep anti-Human, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF5374

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13439224

  • £314.17 / 100µg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PLA2G2A Polyclonal antibody AF5374 Specifically detects PLA2G2A in Human samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

PLA2G2A
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
P14555
EC 3.1.1.4, GIIC sPLA2, Group IIA phospholipase A2, MOM1, Non-pancreatic secretory phospholipase A2, NPS-PLA2, Phosphatidylcholine 2-acylhydrolase 2A, phospholipase A2, group IIA (platelets, synovial fluid), phospholipase A2, membrane associated, PLA2B, PLA2BPLA2, PLA2L, PLA2LPLA2S, PLAS1, RASF-A, sPLA2
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human PLA2G2A in direct ELISAs and Western blots. In Western blots, approximately 60% cross-reactivity with recombinant mouse PLA2G2A is observed and less than 5% cross-reactivity with recombinant human (rh) PLA2G1B and rhPLA2G7 is observed.
Human
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PLA2G2A
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human PLA2G2A Asn21-Cys144 Accession # P14555
Antigen Affinity-purified
RUO
Primary
5320
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only