Learn More
PIN Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF877
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440295
  • £362.60 / 100µg
Estimated Shipment
19-08-2020
Description
Description
PIN Polyclonal antibody AF877 Specifically detects PIN in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
Specifications
PIN/DLC8
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 2.5-5 ug/10^6 cells
P63167
cytoplasmic dynein light polypeptide, DLC1, DLC1DNCLC1, DLC8, DLC8MGC126137, DNCL1, DNCL1MGC126138, DNCLC1, dynein light chain 1, cytoplasmic, Dynein light chain LC8-type 1, dynein, cytoplasmic, light polypeptide 1, dynein, light chain, LC8-type 1,8 kDa dynein light chain, DYNLL1, hdlc1, LC8, LC8a, PINLC8a, Protein inhibitor of neuronal nitric oxide synthase
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human PIN in direct ELISAs and Western blots.
Human
Western Blot, Immunohistochemistry, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
DYNLL1
E. coli-derived recombinant human PIN Trp2-Gly89 Accession # P63167
Antigen Affinity-purified
RUO
Primary
8655
Reconstitute at 0.2 mg/mL in sterile PBS.