missing translation for 'onlineSavingsMsg'
Learn More

pIgR Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2717

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13487734

  • £377.53 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

pIgR Polyclonal antibody AF2717 specifically detects pIgR in Human samples. It is validated for Blocking (Receptor-ligand), Western Blotting
Specifications

Specifications

pIgR
LYOPH
Western Blot 0.2 ug/mL, Blockade of Receptor-ligand Interaction
CAA51532
FLJ22667, hepatocellular carcinoma associated protein TB6, Hepatocellular carcinoma-associated protein TB6, MGC125361, MGC125362, PIgR, Poly-Ig receptor, polymeric immunoglobulin receptor
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human pIgR in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant mouse pIgR is observed.
Human
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PIGR
Mouse myeloma cell line NS0-derived recombinant human pIgR (R&D Systems, Catalog # 2717-PG), Lys19-Arg638, Accession # CAA51532
Antigen Affinity-purified
RUO
Primary
5284
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only