missing translation for 'onlineSavingsMsg'
Learn More

PER1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4565

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13488794

  • £347.17 / 100µg


Estimated Shipment
03-10-2018
Add to basket

Description

Description

PER1 Polyclonal antibody AF4565 Specifically detects PER1 in Human samples. It is validated for Immunohistochemistry
Specifications

Specifications

PER1
LYOPH
Immunohistochemistry 5-15 ug/mL
O15534
Circadian clock protein PERIOD 1, Circadian pacemaker protein Rigui, hPER1, KIAA0482, PERhPER, period (Drosophila) homolog 1, period circadian protein homolog 1, period homolog 1 (Drosophila), Period, drosophila, homolog of, PERIOD1, RIGUI, RIGUIMGC88021
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects human PER1 in direct ELISAs.
Human
Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PER1
E. coli-derived recombinant human PER1, Ser159-Thr365, Accession # O15534
Antigen Affinity-purified
RUO
Primary
5187
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only