Learn More
Peptidylglycine alpha-Amidating Monooxygenase/PAM Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF4837
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458934
  • £389.41 / 100µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
Peptidylglycine alpha-Amidating Monooxygenase/PAM Polyclonal antibody AF4837 Specifically detects Peptidylglycine alpha-Amidating Monooxygenase/PAM in Human samples. It is validated for Western Blotting
Specifications
Specifications
Peptidylglycine alpha-Amidating Monooxygenase/PAM
LYOPH
Western Blot 0.1 ug/mL
P19021
PAL, pancreatic peptidylglycine alpha-amidating monooxygenase, peptidyl alpha-amidating enzyme, peptidyl-alpha-hydroxyglycine alpha-amidating lyase, peptidylamidoglycolate lyase, peptidylglycine 2-hydroxylase, peptidyl-glycine alpha-amidating monooxygenase, peptidylglycine alpha-amidating monooxygenase, peptidylglycine alpha-hydroxylating monooxygenase, PHM
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human PAM in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PAM
Mouse myeloma cell line NS0-derived recombinant human PAM Phe21-Val817 Accession # P19021
Immunogen affinity purified
RUO
Primary
5066
Reconstitute at 0.2 mg/mL in sterile PBS.