missing translation for 'onlineSavingsMsg'
Learn More

Pax4 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2614

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447674

  • £433.65 / 100µg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Pax4 Polyclonal antibody AF2614 Specifically detects Pax4 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications

Specifications

Pax4
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
NP_006184
KPD, MGC129960, MODY9paired box protein Pax-4, paired box 4, paired box gene 4, paired domain gene 4
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Pax4 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) Pax3 is observed.
Human
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PAX4
E. coli-derived recombinant human Pax4 Met1-Glu343 Accession # NP_006184
Immunogen affinity purified
RUO
Primary
5078
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only