missing translation for 'onlineSavingsMsg'
Learn More

Pax3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2457

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437574

  • £389.41 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Pax3 Polyclonal antibody AF2457 Specifically detects Pax3 in Human samples. It is validated for Immunocytochemistry, Western Blotting
Specifications

Specifications

Pax3
LYOPH
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL
NP_000429
CDHS, HuP2, HUP2MGC120384, MGC120381, MGC120382, MGC120383, MGC134778, paired box 3, paired box gene 3 (Waardenburg syndrome 1), paired box homeotic gene 3, paired box protein Pax-3, paired domain gene 3, paired domain gene HuP2, Waardenburg syndrome 1, WS1, WS3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects Pax3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Pax4 is observed.
Human
Western Blot, Immunocytochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
PAX3
E. coli-derived recombinant human Pax3 Met1-Ser215 Accession # NP_000429
Immunogen affinity purified
RUO
Primary
5077
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos

For Research Use Only