missing translation for 'onlineSavingsMsg'
Learn More

Osteoadherin/OSAD Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2884

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13457844

  • £410.13 / 100µg

Estimated Shipment date
from Supplier 23-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Osteoadherin/OSAD Polyclonal antibody AF2884 Specifically detects Osteoadherin/OSAD in Human samples. It is validated for Western Blotting
Specifications

Specifications

Osteoadherin/OSAD/OMD
LYOPH
Western Blot 0.1 ug/mL
Q99983.1
Keratan sulfate proteoglycan osteomodulin, KSPG osteomodulin, OMD, OSAD, Osteoadherin, osteoadherin proteoglycan, osteomodulin, SLRR2C, SLRR2Costeoadherin
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Osteoadherin/OSAD in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant mouse Osteoadherin is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
OMD
Mouse myeloma cell line NS0-derived recombinant human Osteoadherin/OSAD Gln21-Glu421 (Phe67Leu) Accession # Q99983.1
Antigen Affinity-purified
RUO
Primary
4958
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only