missing translation for 'onlineSavingsMsg'
Learn More

Opticin Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF3148

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13468004

  • £360.15 / 100µg

Estimated Shipment date
from Supplier 01-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Opticin Polyclonal antibody AF3148 Specifically detects Opticin in Human samples. It is validated for Western Blotting
Specifications

Specifications

Opticin
LYOPH
Western Blot 0.1 ug/mL
Q9UBM4
oculoglycan, Oculogylcan, OPT, OPTC, opticin
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Opticin in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant mouse Opticin is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
OPTC
Mouse myeloma cell line NS0-derived recombinant human Opticin Ala20-Thr332 Accession # Q9UBM4
Immunogen affinity purified
RUO
Primary
26254
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only