missing translation for 'onlineSavingsMsg'
Learn More

Olig3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2356

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13437504

  • £332.22 / 100µg

Estimated Shipment date
from Supplier 23-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Olig3 Polyclonal antibody AF2356 Specifically detects Olig3 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Olig3
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q7RTU3
Bhlhb7, BHLHE20, bHLHe20oligo3, Class B basic helix-loop-helix protein 7, Class E basic helix-loop-helix protein 20, Oligo3, oligodendrocyte transcription factor 3
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Olig3 in direct ELISAs and Western blots. In Western blots, less than 2% cross-reactivity with recombinant human (rh) Olig-1 and rhOlig-2 is observed.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
OLIG3
E. coli-derived recombinant human Olig3 Met1-Lys272 Accession # Q7RTU3
Immunogen affinity purified
RUO
Primary
167826
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only