missing translation for 'onlineSavingsMsg'
Learn More

Olig1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2417

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447544

  • £446.88 / 100µg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Olig1 Polyclonal antibody AF2417 Specifically detects Olig1 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

Olig1
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
BAD93028
bHLHb6, BHLHE21, Class B basic helix-loop-helix protein 6, Class E basic helix-loop-helix protein 21, helix-loop-helix protein, class B, 6, oligo1, oligodendrocyte lineage transcription factor 1, oligodendrocyte transcription factor 1, oligodendrocyte-specific bHLH transcription factor 1
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human Olig1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows less than 5% cross-reactivity with recombinant human (rh) Olig2 and rhOlig3.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
OLIG1
E. coli-derived recombinant human Olig1 Met12-Lys283 Accession # BAD93028
Antigen Affinity-purified
RUO
Primary
116448
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only