Learn More
NRL Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
Brand:  RD Systems (Bio-Techne) AF2945
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13447884
  • £446.88 / 100µg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
HGF Polyclonal antibody AF294 Specifically detects HGF in Human samples. It is validated for Immunohistochemistry, Neutralization, Western Blotting
Specifications
Specifications
NRL
LYOPH
Western Blot 0.1 ug/mL
P54845
D14S46Eneural retina-specific leucine zipper protein, neural retina leucine zipper, NRL-MAF, RP27, RP27neural retinal-specific leucine zipper
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human NRL in direct ELISAs and Western blots.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NRL
E. coli-derived recombinant human NRL Met1-Leu237 Accession # P54845
Immunogen affinity purified
RUO
Primary
4901
Reconstitute at 0.2 mg/mL in sterile PBS.