missing translation for 'onlineSavingsMsg'
Learn More

NrCAM Isoform 3 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2034

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13467294

  • £332.22 / 100µg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NrCAM Isoform 3 Polyclonal antibody AF2034 Specifically detects NrCAM Isoform 3 in Human samples. It is validated for Immunohistochemistry, Western Blotting
Specifications

Specifications

NrCAM
LYOPH
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Q14CA1
bravo, hBravo, KIAA0343Neuronal surface protein Bravo, MGC138845, MGC138846, neuronal cell adhesion molecule, Ng-CAM-related, NgCAM-related cell adhesion molecule, nr-CAM
Goat
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human NrCAM Isoform 3 in direct ELISAs and Western blots.
Human
Western Blot, Immunohistochemistry
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
NRCAM
Mouse myeloma cell line NS0-derived recombinant human NrCAM isoform 3 Leu30-Asn600 (Ala526Pro) Accession # Q14CA1
Immunogen affinity purified
RUO
Primary
4897
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only